Pipette controller, MACROMAN™
Pipette controller, MACROMAN™, 1 - 100 ml
Catalog # GILSF110120
Supplier: GILSON
Jump to:
Product Details & Documents
Pipette controller for all plastic and glass pipettes from 1 to 100 ml.
- Ergonomic handle featuring wide triggers for maximum comfort and stress-free pipetting
- Lithium-Ion battery enables a longer, vibration-free use with reduced charging time
- Increase precision by choosing your flow rate mode from slow drops to rapid dispensing
Documents
Specifications
- Kapacitet:1 - 100 ml
- Beskrivning:Pipette controller, MACROMAN™
Accessories
Product Information
- Catalog #BeskrivningAvailabilityPrice
- GILSF107204Filter, 0,22 µm, standard (set of 10)Log in for availability1 * 10 Items - 1 350,00 kr
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
Product Information
- Catalog #BeskrivningAvailabilityPrice
- GILSF107206Filter, 0,45 µm, standard (set of 10)Log in for availability1 * 10 Items - 1 350,00 kr