Transfection reagent, Pro-DeliverIN™
Leverantör: OZ BIOSCIENCES
Certifikat
Om denna produkt
Pro-DeliverIN™ transfection reagent is an innovative reagent allowing intracellular delivery of biologically active proteins. This lipid-based formulation is serum compatible to deliver biologically functional proteins into living cells.
- Intracellular delivery of functionally active proteins
- Highly efficient in many primary cells and cell lines
- Serum compatible
- Non toxic
- Straightforward protocol and ready to use reagent
Store at 4 °C.
Delivery: Shipped at room temperature. Pro-DeliverIN™ is provided with 100 µl of R-Phycoerythrin positive control.
Specifikationer
- Protein delivery:Ja