Broxettes of evaporated salt tablets
Katalog # KAMM1161131
Leverantör: KAMMERER
Specifikationer
- Beskrivning:Broxettes of evaporated salt tablets
- Förp.:25 kg bag
Specifikationer
Om denna produkt
Pressed evaporated salt tablets for the regeneration of ion exchangers.
- Highest degree of purity
- Tested and suitable for chlorine chlorinators (according to EN 973 type A)