Disposable syringes, Norm-Ject®
Katalog # MEGRHSW4200.090D0
Leverantör: MEGRO
Specifikationer
- Kapacitet:20 ml
- Beskrivning:Syringe, 2-parts
- Seq:02
- Förp.:1.000
Specifikationer
Om denna produkt
Standard-disposable syringe, easy working and absolutely leak-proof, clear as crystal cylinder, scale is easy to read, with a second braking ring on the bottom of the cylinder.
- Piston is made of polyethylene
- Cylinder is made of polypropylene
Second braking ring on the bottom of the cylinder. thereby the resistance will be increased to separate the piston and the cylinder, free of latex and silicone oil.