XK and K 9 series O-rings
19-0685-01
: Cytiva
- Beskrivning:O-Ring for use with XK 50 columns, 25,07×2,62 mm
- Material:NBR
- Seq:10
- Förp.:5
Can be used in aqueous solutions and nearly all organic solvents commonly used in liquid chromatography of macromolecules with the following few exceptions - chlorinated hydrocarbons, acetone and other ketones, aliphatic esters and phenol
Solutions containing more than 10% NaOH, 10% HCl, other strong mineral acids or 5% acetic acid should not be used.