Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Kolorimetrar
Komparatorkit
Accessories for Nessleriser systems
Accessories for Nessleriser systems
Accessories for Nessleriser systems
Catalog # TINT354230
Supplier:
Lovibond Tintometer
Expand image
Accessories for Nessleriser systems
Catalog # TINT354230
Supplier:
Lovibond Tintometer
Specifications
Beskrivning:
DB423 pair of Nessler tubes, 100 ml/288 mm (ASTM D 1209) with glass anti-meniscus plungers
Förp.:
2
Specifications
About this item
Product Details
Product Details