Yeast extract, Veggie™
Catalog # 71279-3
Supplier: Merck Millipore (Novagen)
CAS Number:
Specifications
- Förp.:500 g
- Beskrivning:Veggie™ yeast extract
Specifications
About this item
Veggie yeast extract is a certified animal-free media component.
- Ensures proper growth and maintenance of bacterial cells
- Ideal for applications requiring minimal use of animal-derived materials
Used as a direct replacement for traditional yeast extract in bacterial growth media.