Hydrophobic interaction chromatography columns, HiTrap™ Capto™ Phenyl (High Sub)
Leverantör: Cytiva
Certifikat
Om denna produkt
Capto™ Phenyl (High sub) and Capto Butyl are hydrophobic interaction chromatography (HIC) resins used in the capture and intermediate stages of protein purification.
- Aromatic ligand coupled with a flexible spacer for laboratory and process applications requiring a HIC resin with resin to high hydrophobicity
- Highly rigid matrix enables high flow rates, long bed heights and faster throughput
- Improves productivity and process economy in downstream operations
- Narrow specification range ensures robust performance in sensitive processes
- Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide
Specifikationer
- Catalog No:
- 17-5451-08
- GEHE17-5451-09
- Förvaring:
- 4 to 30°C, 20% Ethanol+
- 4 to 30°C, 20% Ethanol+
Specifikationer
Relaterad information
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements