Fenantren
Glass bottle for solids, 100 g
Katalog # MOLE17348375-100G
Marketsource
Leverantör: Molekula
Gå till:
Product Details & Documents
- CAS-nummer:85-01-8
- Formel:C₁₄H₁₀
- MDL-nummer:MFCD00001168
- Molecular Weight:178.23 g/mol
- Merck Index:12,07354
- Kokande Pt:340 °C (1013 hPa)
- Smält Pt:97…99 °C
- Densitet:1,19 g/cm³ (20 °C)
- Flash Pt:153 °C
- Förvaringstemperatur:Omgivande
- EINECS:201-581-5
- FN:3077
- ADR:9,III
Specifikationer
Specifikationer
- Förpackningstyp:Glass bottle for solids
- Förp.:100 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



