Order Entry
Sweden
ContactUsLinkComponent
Silversulfat ≥99%, Puriss., Fluka™
Corrosive
Silversulfat ≥99%, Puriss., Fluka™
Catalog # 10229-500G
CAS Number:  
undefined
Corrosive
Silversulfat ≥99%, Puriss., Fluka™
Catalog # 10229-500G
CAS Number:  

Specifications

  • Förpackningstyp:
    Plastic bottle
  • Förp.:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • Assay:
    Min. 99 %
  • Copper (Cu):
    Max. 0.005 %
  • Iron (Fe):
    Max. 0.005 %
  • Lead (Pb):
    Max. 0.005 %
  • Chloride (Cl):
    Max. 0.001 %
  • Nitrate (NO₃):
    Max. 0.01 %

Specification Test Results

About this item

 :
 :311.8 g/mol
Merck Index:13,08603
Boiling Pt:Decomposition
Boiling Pt: Dec.
Melting Pt:655 °C
Density:5,45 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:233-653-7
 :GHS05
CAS Number:10294-26-5
Formula:Ag₂SO₄
MDL Number:MFCD00003407