Order Entry
Sweden
ContactUsLinkComponent
Ammoniumcerium (IV) nitrat
Oxidizing
Corrosive
Irritant
Ammoniumcerium (IV) nitrat
Catalog # GFSC13
Supplier:  GFS CHEMICALS
CAS Number:  
Oxidizing
Corrosive
Irritant
Ammoniumcerium (IV) nitrat
Catalog # GFSC13
Supplier:  GFS CHEMICALS
CAS Number:  

Specifications

  • Förp.:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :548.22 g/mol
Merck Index:13,00509
Melting Pt:107…108 °C
Density:2,2 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:240-827-6
UN:1477
ADR:5.1,III
 :GHS03
 :GHS05
 :GHS07
CAS Number:16774-21-3
Formula:(NH₄)₂Ce(NO₃)₆
MDL Number:MFCD00151121