ContactUsLinkComponent
Dispenser tips
Dispenser tips
Katalog # 612-6612
Leverantör:  Alpha Laboratories
Dispenser tips
Katalog # 612-6612
Leverantör:  Alpha Laboratories
Specifikationer
  • Förp.:
    Loose
  • Steril:
    -
  • Volym:
    1,25 ml
  • Seq:
    01
  • För användning med:
    Ripette® Stepper
Specifikationer
Om denna produkt
This dispenser tip is designed for repetitive dosing application. Tip graduated with 0,25 ml increments and constructed with a polypropylene barrel and a polyethylene piston.
It is also compatible with the Sartorius Biohit Manual Dispenser, Eppendorf Multipette® 4780, Gilson Distriman®, HTL Minilab 100/101 and the Brand HandyStep®.