Order Entry
Sweden
ContactUsLinkComponent
 
Etyltriklorsilan för syntes, Sigma-Aldrich®
Catalog # 8.21667.0025
Supplier:  Merck
CAS Number:  
Etyltriklorsilan för syntes, Sigma-Aldrich®
Catalog # 8.21667.0025
Supplier:  Merck
CAS Number:  

Specifications

  • Förp.:
    25 mL
  • Seq:
    2315

Specifications

Specification Test Results

  • Solubility:
    (20 °C) (decomposition)
  • Melting Point:
    -105 °C
  • Molar Mass:
    163.5 g/mol
  • Boiling Point:
    98 °C (1013 hPa)
  • Vapor Pressure:
    36 hPa (20 °C)
  • Flash Point:
    14 °C
  • Refractive Index:
    1.4257 (20 °C, 589 nm)
  • Explosion Limit:
    2.68 %(V)
  • Density:
    1.24 g/cm3 (20 °C)
  • Ignition Point:
    385 °C

Specification Test Results

About this item

 :
 :163.51 g/mol
Boiling Pt:102 °C (1013 hPa)
Density:1,24 g/cm³ (20 °C)
ADR:3 (8),II
 :M-Clarity™ quality level = MQ200
EINECS:204-072-6
UN:1196
 :GHS02
 :GHS05
 :GHS07
CAS Number:115-21-9
Formula:C₂H₅Cl₃Si
MDL Number:MFCD00000485