Natriumhydroxid
25 kg
Katalog # BREN5268
Marketsource
Leverantör: BRENNTAG
Gå till:
Product Details & Documents
Synonymer: Natronlut, Kaustik soda
- CAS-nummer:1310-73-2
- Formel:NaOH
- MDL-nummer:MFCD00003548
- Molecular Weight:40 g/mol
- Merck Index:14,08627
- Kokande Pt:1390 °C (1013 hPa)
- Smält Pt:323 °C
- Densitet:2,13 g/cm³ (20 °C)
- Förvaringstemperatur:Omgivande
- EINECS:215-185-5
- FN:1823
- ADR:8,II
- REACH:01-2119457892-27
Specifikationer
Specifikationer
- Förp.:25 kg
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



