Fenolrött
Catalog # MOLEM86776424
Supplier: Molekula
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:100 g
- Seq:3401
Specifications
About this item
:
Merck Index:13,07329
Melting Pt:> 300 °C
:354.38 g/mol
Storage Temperature:Omgivande
EINECS:205-609-7
CAS Number:143-74-8
Formula:C₁₉H₁₄O₅S
MDL Number:MFCD00003552