2-Metyl-2-butanol, EMPLURA®, Supelco®
Catalog # 8.06193.1000
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:1 L
- Seq:5037
Specifications
About this item
:
:88.15 g/mol
Merck Index:13,07218
Boiling Pt:102 °C (1013 hPa)
Density:0,806 g/cm³ (20 °C)
Flash Pt:20 °C
Storage Temperature:Omgivande
EINECS:200-908-9
UN:1105
ADR:3,II
:M-Clarity™ quality level = MQ200
:GHS02
:GHS05
:GHS07
CAS Number:75-85-4
Formula:C₅H₁₂O
MDL Number:MFCD00004478