Tris-(hydroximetyl) aminometan, Millipore®
Catalog # 648310-2.5
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:2,5 kg
- Seq:4076
Specifications
About this item
:
:121.14 g/mol
Merck Index:13,09842
Boiling Pt:357 °C (1013 hPa)
Melting Pt:169…172 °C
Density:1,328 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:201-064-4
:GHS07
CAS Number:77-86-1
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679