


Methyl trifluoromethanesulphonate 96%
Catalog # ACRO415011000
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:100 g
- Seq:3401
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3250 to 1.3270 (20°C, 589 nm)
- GC:≥95 %
- Enantiomeric excess:≥99.0 % (HPLC)
Specification Test Results
About this item
:
:164.11 g/mol
Boiling Pt:95 °C (760 mmHg)
Density:1,45 g/cm³
Flash Pt:38 °C
Storage Temperature:Kylskåp
EINECS:206-371-7
UN:2920
ADR:8,II
:GHS07
:GHS05
:GHS02
CAS Number:333-27-7
Formula:C₂H₃F₃O₃S
MDL Number:MFCD00000409