

Glykolsyra, EMSURE® Reag. Ph. Eur. for analysis, Supelco®
Catalog # 1.04106.0100
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Plastic bottle
- Förp.:100 g
- Seq:3401
Specifications
Specification Test Results
- Assay (acidimetric):≥ 99.5 %
- Identity (IR):passes test
- Melting range (lower value):≥ 75 °C
- Melting range (upper value):≤ 79 °C
- Heavy metals (as Pb):≤ 10 ppm
- Iron (Fe):≤ 5 ppm
- Water (K. F.):≤ 0.50 %
- Density:1.49 g/cm3 (25 °C)
- pH:2 (50 g/l, H2O, 20 °C)
Specification Test Results
About this item
:
:76.05 g/mol
Merck Index:13,04511
Boiling Pt:266 °C (1013 hPa)
Melting Pt:79…80 °C
Density:1,49 g/cm³ (20 °C)
Flash Pt:> 300 °C
EINECS:201-180-5
UN:3261
ADR:8,II
:M-Clarity™ quality level = MQ300
:GHS05
:GHS07
Storage Temperature:Omgivande
CAS Number:79-14-1
Formula:C₂H₄O₃
MDL Number:MFCD00004312