Ammoniumformat 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:540-69-2
:
Formula:HCO₂NH₄
MDL Number:MFCD00013103
:63.06 g/mol
Merck Index:13,00525
Melting Pt:115…120 °C
Density:1,28 g/cm³ (25 °C)
EINECS:208-753-9
:GHS07
Storage Temperature:Omgivande
Specification Test Results
- Appearance:White Powder or crystals
- Infrared spectrum:Conforms
- Titration with HClO4:96.0 to 104.0 %
- HPLC:≥94.0 %