Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Standarder
Anionic Standards
Phosphate Standards
Phosphate standard for BIOMOL® GREEN
Phosphate standard for BIOMOL® GREEN
Phosphate standard for BIOMOL® GREEN
Katalog # ENZOBMLKI1020001
Leverantör:
ENZO LIFE SCIENCES
CAS-nummer:
Expandera bilden
Phosphate standard for BIOMOL® GREEN
Katalog # ENZOBMLKI1020001
Leverantör:
ENZO LIFE SCIENCES
CAS-nummer:
Specifikationer
Koncentration:
800 µM
Förp.:
1 mL
Beskrivning:
Phosphate standard for BIOMOL® green
Seq:
02