DNA ladder 50 to 1500 bp, AMPIGENE®
Katalog # ENZOENZGEN1020100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Förp.:1 SET
- Beskrivning:AMPIGENE® 50 - 1500 bp ladder, sufficient for 100 lanes
- Seq:02
Specifications
About this item
AMPIGENE® DNA ladder contains DNA fragments ranging from 50 bp to 1500 bp that can be used as molecular weight standards for agarose gel electrophoresis. The bands are evenly spaced and are intensified at 200 bp, 500 bp and 1500 bpto serve as easily identifiable reference bands. The approximate mass of DNA in each of the bands is provided, assuming 5 µl load, for determining the approximate mass of DNA in samples with comparable intensity and sizes.
- Ready to use format can be loaded straight onto a gel
- Evenly spaced bands with intensified reference bands
- Stable at room temperature
Delivery: Kit contains ready to use AMPIGENE® DNA ladder 50 – 1500 bp and sample loading buffer.