Plate rack, ImmunoSet®
Katalog # ENZOADI801930
Leverantör: ENZO LIFE SCIENCES
Specifikationer
- Ant. brunnar:96
- Färg:Klar
- Beskrivning:IMMUNOSET® plate rack
- Seq:02
- Förp.:1
Specifikationer
Om denna produkt
This ImmunoSet® plate rack is a convenient set of five 96 well, clear, high-binding, polystyrene plates and 5 plate sealers.
Use of these plates will provide optimal performance. The use of different types of plates will produce different results and may affect the sensitivity of the assay due to the type of plastic used.