Magnesiumsulfat heptahydrat, Supelco®
Catalog # 1.11047.9011
Supplier: MERCK PRODUCTION CHEMICALS
CAS Number:
Specifications
- Förp.:10 kg
- Seq:4158
Specifications
About this item
:
:246.48 g/mol
Merck Index:13,05715
Melting Pt:1124 °C
Density:2,66 g/cm³ (20 °C)
CAS Number:10034-99-8
Formula:MgSO₄·7H₂O
MDL Number:MFCD00149785