DL-leucin
Catalog # EHERC14629300
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Förpackningstyp:Glass bottle for solids
- Förp.:0,1 g
- Seq:3020
Specifications
About this item
:
:131.17 g/mol
Merck Index:13,05470
Boiling Pt:226 °C (1013 hPa)
Density:1,191 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:206-328-2
CAS Number:328-39-2
Formula:C₆H₁₃NO₂
MDL Number:MFCD00063087