Nitrile gloves, Dexpure® 800-91
Supplier: HONEYWELL SAFETY
Certificates
About this item
These disposable nitrile gloves offer better mechanical resistance than latex gloves.
- Lightly powdered, non sterile
- Patterned finish provides a better grip
- Powdered interior facilitates putting the glove on and taking it of
- Treated to prevent the development of bacteria
- Blue colour: Glove is easy to detect, which makes it suitable for the agrifood industry
- Good alternative for workers allergic to natural latex
Typical applications include: Agrifood industry, electronics and complex assembly, graphic industry, painting in the automotive industry, pharmaceutical laboratories, chemicals.
Certifications: EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS); Cat. II
Food contact approved
Food contact approved
Specifications
- Catalog No:
- DALL4580091/L
- DALL4580091/M
- DALL4580091/S
- DALL4580091/XL
- Ej handspecifika/handspecifika:
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Latex/latexfri:
- Latexfria
- Latexfria
- Latexfria
- Latexfria
- Pudrad/puderfri:
- Lätt pudrade
- Lätt pudrade
- Lätt pudrade
- Lätt pudrade
- Yta:
- Textured
- Textured
- Textured
- Textured
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- EN standard:
- EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)
- EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)
- EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)
- EN ISO 374-1:2016 Type C, EN ISO 374-5:2016 (MICRO-ORGANISMS)
- Godkända för livsmedelskontakt:
- +
- +
- +
- +