Deuteriumoxid ≥99.95%, MagniSolv™ for NMR spectroscopy pH 7 (H₂O, 20 °C), Sigma-Aldrich®
Catalog # 1.03428.0100
Supplier: Merck
CAS Number:
Specifications
- Förpackningstyp:Glass bottle
- Förp.:100 mL
- Seq:2440
Specifications
Specification Test Results
- Deuteration degree:≥ 99.95 %
- Performance test (NMR-spectrum):passes test
Specification Test Results
About this item
:
:20 g/mol
Merck Index:12,02984
Boiling Pt:101,3 °C (1013 hPa)
Density:1,1 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Kylskåp
EINECS:232-148-9
CAS Number:7789-20-0
Formula:D₂O
MDL Number:MFCD00044636