Mercury short arc reflector lamp
Catalog # ZEIS0000001313162
Supplier: ZEISS
Specifications
- För:X-Cite 120 illuminator
- Beskrivning:Mercury short arc reflactor lamp module
- Förp.:1
Specifications
About this item
This mercury short arc reflector lamp is specially designed for X-Cite 120 illuminator system. The average lifetime performance of this lamp module is 2000 hours.