Molekylsikt A4, (0,4 nm, 4 Å), pellets, Ø 3.2 mm, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:70955-01-0
:1318-02-1
MDL Number:MFCD00131613
Melting Pt:400 °C
Density:2,0 g/cm³
Storage Temperature:Omgivande
Specification Test Results
- Diameter:3.2 mm
- Form:pellets
- Granulation:complying
- Pore Size (Ång Ström):4