Molekylsikt 13x (1,0 nm, 10 Å), beads 8-12 mesh, Fluka™
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:63231-69-6
:1318-02-1
MDL Number:MFCD00131613
Storage Temperature:Omgivande
Specification Test Results
- Form:beads
- Grain size 8 - 12 mesh:Min. 95 %