ContactUsLinkComponent
Aluminium ≥99.0%, thin foil, Thickness: 10.0 µm (0.0004 in)
Catalog # 41798.GH
CAS Number:  
Aluminium ≥99.0%, thin foil, Thickness: 10.0 µm (0.0004 in)
Catalog # 41798.GH
CAS Number:  
Specifications
  • Förp.:
    1
  • Seq:
    1736
  • Pk Info:
    100×100 mm
Specifications
About this item
CAS Number:7429-90-5
 :
Formula:Al
MDL Number:MFCD00134029
 :26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2,7 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:231-072-3