Aluminium ≥99.99%, tråd, hard, Ø 0.025 mm (0.001 in)
Catalog # 40677.G1
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Förp.:1 m
- Seq:8362
Specifications
About this item
CAS Number:7429-90-5
:
Formula:Al
MDL Number:MFCD00134029
:26.98 g/mol
Merck Index:13,00321
Boiling Pt:2327 °C (1013 hPa)
Melting Pt:660 °C
Density:2,7 g/cm³ (20 °C)
Storage Temperature:Omgivande
EINECS:231-072-3