ContactUsLinkComponent
Culture media, in Petri dishes, Slanetz Bartley medium
Culture media, in Petri dishes, Slanetz Bartley medium
Catalog # APPC443812.0922
Supplier:  PanReac AppliChem
CAS Number:  
undefined
Culture media, in Petri dishes, Slanetz Bartley medium
Catalog # APPC443812.0922
Supplier:  PanReac AppliChem
CAS Number:  
Specifications
  • Förp.:
    30
  • Beskrivning:
    Slanetz Bartley medium, plate Ø 55 mm
Specifications
About this item
Prepared medium for the enumeration of Enterococci.
  • Pinkish beige