Cardiotoxicity library, Screen-Well®
Leverantör: ENZO LIFE SCIENCES
Certifikat
Om denna produkt
The Screen-Well® cardiotoxicity library is a focused collection of 130 compounds with defined and diverse cardiotoxicity, including ion channel blockage, mitochondrial toxicity, arrhythmia, fibrosis, and many more. A variety of structurally and mechanistically different compound classes are included, as well as nontoxic controls. The compounds are dissolved in DMSO at 10 mM and aliquoted into deep-well plates. The library is an essential tool for predictive toxicology screening and assay development.
Ordering information: Contains 130 compounds with defined and diverse cardiotoxicity. A variety of structurally and mechanistically different compound classes, as well as nontoxic controls, are included.
Specifikationer
- Koncentration:DMSO solutions (10 mM )