Order Entry
Sweden
ContactUsLinkComponent
Z-AAD-CMK
Z-AAD-CMK
Katalog # ENZOBMLP1650001
Leverantör:  ENZO LIFE SCIENCES
CAS-nummer:  
Z-AAD-CMK
Katalog # ENZOBMLP1650001
Leverantör:  ENZO LIFE SCIENCES
CAS-nummer:  

Specifikationer

  • Förp.:
    1 mg
  • Seq:
    02
  • Protein/peptide name:
    Z-AAD-CMK
  • Renhet:
    >95% (HPLC)

Specifikationer

Om denna produkt

Caspase inhibitor

Irreversible, cell-permeable inhibitor of granzyme B (ID50=0,3 µM). Does not inhibit granzyme A. Inhibition of granzyme B with Z-AAD-CMK prevented caspase-3 activation in T cells.