Protein tyrosine phosphatase substrate
Catalog # ENZOBMLP1530001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Förp.:1 mg
- Seq:02
- Protein/peptide name:Protein tyrosine phosphatase substrate
Specifications
About this item
This peptide sequence, ENDpYINASL, is derived from a highly conserved region of the T-cell phosphatase, TCPTP, and has been shown to be an excellent general substrate for protein tyrosine phosphatases.