Protein phosphatase 2 substrate
Katalog # ENZOBMLP1520001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Förp.:1 mg
- Seq:02
- Protein/peptide name:Protein phosphatase 2 substrate
Specifications
About this item
This peptide sequence, RRApTVA, has been found to be an excellent substrate for both PP2A and PP2C phosphatases.