Rat ribosomal protein S6 (229-239)
Catalog # ENZOBMLP1440001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Förp.:1 mg
- Species:Rat
- Seq:02
- Protein/peptide name:Ribosomal protein S6 (229-239)
Specifications
About this item
S6 kinase substrate
This sequence, AKRRRLSSLRA, derived from rat hepatic 40S ribosomal protein S6 (229-239). Substrate for Rho-kinase II (ROCK II), Km=0,9 µM and for p90 rsk (S6K) in cell extracts (Km=0,18 mM) using phosphocellulose assay. Peptide phosphorylation by PKS and PKC can be eliminated by the inclusion of PKI and EGTA, respectively, in the assay.