Human MMP8 proenzyme (from neutrophils)
Catalog # ENZOALX200421C005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Förp.:5 µG
- Källa:Neutrophils
- Species:Human
- Seq:02
- Protein/peptide name:MMP8 proenzyme
- Renhet:>90% (SDS-PAGE)
- Koncentration:100 µg/ml
- Formulering:Liquid. In 50 mM TRIS-HCl, pH 7,0, containing 200 mM NaCl, 5 mM CaCl₂, 1 µM ZnCl₂, 0,05% BRIJ 35, and 0,05% sodium azide.
Specifications
About this item
MMP-8 is a glycoprotein containing complex N-linked oligosaccharides.
- Hydrolyzes type I over type II, and III collagens
- Activated MMP-8 is inhibited by TIMP-1 (Prod. No. ALX-200-426)
Isolated from stimulated human neutrophil granulocytes (buffy coat). Requires activation.