Order Entry
Sweden
ContactUsLinkComponent
 
Mouse;Rat guanylin
  ENZOALX157011MC01
 :  
Mouse;Rat guanylin
  ENZOALX157011MC01
 :  

 

  • Förp.:
    0,1 mg
  • Species:
    Rat
    Mouse
  • Seq:
    02
  • Protein/peptide name:
    Guanylin
  • Renhet:
    >99%
  • Formulering:
    Lyophilised

 

 

Guanylyl cyclase activator

Endogenous intestinal guanylyl cyclase activator.