EpiXtract® Nuclear protein isolation kit
ENZOENZ450160100
:
- Förp.:1 KIT
- Beskrivning:EpiXtract® nuclear protein isolation kit
- Seq:02
Rapid, reliable method to isolate nuclear proteins in mammalian cells and tissues.
For the extraction of nuclear proteins from mammalian cells or tissues.
EpiXtract® Nuclear Protein Isolation Kit II provides a simple and reliable method to isolate nuclear proteins. Extracts can be used for a variety of downstream applications including western blotting, protein-DNA binding assays, nuclear enzyme assays and others requiring nuclear proteins.
: Contains Pre-extraction Buffer, Extraction Buffer, Protease Inhibitor Cocktail, DTT.