EpiXtract® Nuclear protein isolation kit II (nucleic acid-free)
Katalog # ENZOENZ450150100
Leverantör: ENZO LIFE SCIENCES
CAS-nummer:
Specifikationer
- Förp.:1 KIT
- Beskrivning:EpiXtract® nuclear protein isolation kit II (nucleic acid-free)
- Seq:02
Specifikationer
Om denna produkt
Rapid, reliable method to isolate nucleic acid-free nuclear proteins in mammalian cells and tissues.
For the extraction of nucleic acid-free nuclear proteins from mammalian cells or tissues.
EpiXtract® Nuclear Protein Isolation Kit II provides a simple and reliable method to isolate nucleic acid-free nuclear proteins. Extracts can be used for a variety of downstream applications including western blotting, protein-DNA binding assays, nuclear enzyme assays and others requiring nucleic acid-free nuclear proteins.
Ordering information: Contains Pre-extraction Buffer, Extraction Buffer, Extraction pre-cleaner, Extraction cleaner, Protease Inhibitor Cocktail, DTT.