PROTEOSTAT® thermal shift stability assay kit
Leverantör: ENZO LIFE SCIENCES
Certifikat
Om denna produkt
This kit has been designed for monitoring protein stability under systematic thermal stress conditions.
- Accelerated screening for protein stability as a function of pH, ionic strength, and concentration
- Can detect ligand binding, even without a prior knowledge of the protein’s function or ligand binding site
- Provides a convenient, complementary orthogonal method for cross-validation of instrumentation-intensive techniques
The aggregation temperature is an indicator of protein stability and can be used to optimize conditions that minimize protein aggregation as well as to identify ligands that bind and confer structural stability to a protein of interest. Conditions that increase the aggregation temperature, increase the stability of the protein.
Delivery: Kit includes PROTEOSTAT® TS Detection Reagent, β-Lactoglobulin control, 10X Assay Buffer.
Caution: RUO - Research Use Only.
Specifikationer
- Storage conditions:–20 °C (Short term), –80 °C (Long term)
- Storlek:400 tests