PKC kinase activity kit
Catalog # ENZOADIEKS420A
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Förp.:1 KIT
- Beskrivning:PKC kinase activity kit
- Seq:02
- Storlek:96 wells
Specifications
About this item
The PKC Kinase activity kit is an efficient assay that utilizes little sample volume. This kit gives you the ability to an end-point or kinetic assay read-out in a convenient 96-well plate based assay and offers easy sample handling protocols. The assay also offers a high signal to background ratio.
For the measurement of PKC kinase activity partially purified, purified, or crude enzyme preparations from any species. Cited sample type includes tissue (ref. 1).
Ordering information: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2