IL-10 (human) ELISA Kit
IL-10 (human), ELISA kit, 1×96 wells, 1 KIT
ENZOADI900036
The IL-10 (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit kor the quantitative determination of human IL-10 in culture supernatants, plasma, and serum.
- Ready to use, liquid colour coded reagents and pre-coated plates reduce errors and save time
- Sensitive - detect <4 pg/ml of IL-10
- Results in <3 hours from up to 39 samples in duplicate
: Kit includes microtiter plate, conjugate, antibody, assay buffer 13 concentrate, wash buffer concentrate, standard, TMB substrate, stop solution 2.
- Förp.:1 KIT
- Beskrivning:IL-10 (human), ELISA kit, 1×96 wells
- Förvaringsförhållanden:4 °C (long term), –20 °C (for standards)
- Cat. No.:ENZOADI900036
- Känslighet:3,75 pg/ml (range 7,81 - 500 pg/ml)
- Primary antibody reactivity:Human
- Target protein:Interleukin 10 (IL-10)