Endothelin-1 ELISA kit
ENZOADI900020A
:
- Förp.:1 KIT
- Target protein:Endothelin-1
- Beskrivning:Endothelin-1, ELISA kit, 96 wells
- Seq:02
- Storage conditions:4 °C (long term)
- Detektionsmetod:Colorimetric
- Känslighet:0,40 pg/ml (0,78 - 100 pg/ml)
The Endothelin-1 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 2 hours. This ultra-sensitive kit lets you quantify as little as 0.41 pg/mL ET-1 in a variety of biological matrices with the option of overnight or a 1 hour primary incubation.
- Highly sensitive measurement
- Higher throughput format
- Flexible option of overnight or 1 hour primary incubation
- Suitable for a variety of sample matrices
For the quantitative determination of ET-1 in culture supernatants, plasma, and serum of human, mouse, rat, cow, dog, pig, and rabbit origin.
: Kit contains microtiter plate, standard, antibody, conjugate, assay buffer 28, TMB substrate, stop solution 2, wash buffer concentrate.