Order Entry
Sweden
ContactUsLinkComponent
Anti-SHC1 Mouse Monoclonal Antibody [clone: 11F6]
Anti-SHC1 Mouse Monoclonal Antibody [clone: 11F6]
Catalog # ENZOADI905665100
Supplier:  ENZO LIFE SCIENCES
Anti-SHC1 Mouse Monoclonal Antibody [clone: 11F6]
Catalog # ENZOADI905665100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    SHC-transforming Protein 1
  • Antigensymbol:
    SHC1
  • Klonalitet:
    Monoclonal
  • Klon:
    11F6
  • Konjugation:
    Unconjugated
  • ELISA:
    Yes
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • Isotyp:
    IgG1
  • Reaktivitet:
    Human,
    Mouse
  • Western blot:
    Yes
  • Seq:
    02
  • Epitop:
    C-Terminal
  • Form:
    Lyophilized
  • Antigen synonymer:
    SHC|SH2 domain protein C1|FLJ26504|SHC1|p66SHC|Src homology 2 domain-containing-transforming protein C1|SHC-transforming protein 1|p52SHC|p66|SHCA
  • Storage buffer:
    PBS containing 0,09% sodium azide, PEG, and sucrose.
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Synthetic peptide corresponding to the sequence near the C-terminus of Shc.
  • Rening:
    purified by thiophilic adsorption and size exclusion chromatography
  • Förp.:
    100 µG

Specifications

About this item

The Src homology 2 domain containing (Shc) adaptor protein links growth factor, cytokine, and antigen receptor stimulation to the Ras signaling pathway, and plays a critical role in insulin signaling, tumorigenesis, and hematopoietic cell growth and differentiation. Structurally, Shc is composed of an N-terminal phosphotyrosine binding (PTB) domain, a medial CH1 domain, and a C-terminal SH2 domain. Upon binding to activate cell-surface receptors, Shc is phosphorylated at three tyrosine residues (Tyr239, Tyr240, Tyr317) triggering recognition of Shc by SH2-containing proteins such as Grb2.

Type: Primary
Antigen: SHC1
Clonality: Monoclonal
Clone: 11F6
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse