Order Entry
Sweden
ContactUsLinkComponent
Anti-BAD Mouse Monoclonal Antibody [clone: 5E6]
Anti-BAD Mouse Monoclonal Antibody [clone: 5E6]
Catalog # ENZOADI905638100
Supplier:  ENZO LIFE SCIENCES
Anti-BAD Mouse Monoclonal Antibody [clone: 5E6]
Catalog # ENZOADI905638100
Supplier:  ENZO LIFE SCIENCES

Specifications

  • Antikroppstyp:
    Primär
  • Antikroppsnamn:
    BCL2-associated agonist of cell death
  • Antigensymbol:
    BAD
  • Klonalitet:
    Monoclonal
  • Klon:
    5E6
  • Konjugation:
    Unconjugated
  • Värd:
    Mouse
  • ImmunKemi:
    Yes
  • Immunutfällning:
    Yes
  • Isotyp:
    IgG1
  • Reaktivitet:
    Human
  • Western blot:
    Yes
  • Seq:
    02
  • Form:
    Lyophilized
  • Antigen synonymer:
    BCL2L8|Bcl-2-binding component 6|BBC6|Bcl2 antagonist of cell death|BBC2|BAD
  • Storage buffer:
    PBS containing 0,09% sodium azide, PEG, and sucrose.
  • Förvaringstemperatur:
    –20 °C
  • Shipping temperature:
    Shipped on blue ice
  • Immunogen:
    Recombinant human Bad.
  • Rening:
    purified by thiophilic adsorption and size exclusion chromatography
  • Förp.:
    100 µG

Specifications

About this item

Bad is an ~22 kDa pro-apoptotic protein that aids in the initiation of cell death. It exhibits sequence homology to Bcl-2, a protein that prevents apoptosis, within its BH1 and BH2 domains. Bad is phosphorylated at several sites including Ser112, Ser136; phosphorylation by kinases like Akt is known to alter the effects of Bad. Bad has been demonstrated to be phosphorylated at Ser170.

Type: Primary
Antigen: BAD
Clonality: Monoclonal
Clone: 5E6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human