ContactUsLinkComponent
Anti-CLIP Mouse Monoclonal Antibody
Anti-CLIP Mouse Monoclonal Antibody
Katalog # ENZOADICSA600F
Leverantör:  ENZO LIFE SCIENCES
Anti-CLIP Mouse Monoclonal Antibody
Katalog # ENZOADICSA600F
Leverantör:  ENZO LIFE SCIENCES
Specifikationer
  • Antikroppstyp:
    Primär
  • Antigensymbol:
    CLIP
  • Klonalitet:
    Monoclonal
  • Flödescytometri:
    Yes
  • Värd:
    Mouse
  • Immunutfällning:
    Yes
  • Isotyp:
    IgG1
  • Reaktivitet:
    Human
  • Seq:
    03
  • Storlek:
    200 µg
  • Förp.:
    200 µG
Specifikationer
Om denna produkt
CLIP (class II-associated invariant chain peptide) is an important peptide in the MHC-II antigen processing pathway, formed by the enzymatic cleavage of amino acids 81-104 from the invariant chain. During MHC-II synthesis in the ER and transport to endocytic vesicles, the antigen binding site of MHC-II is obstructed by the presence of the invariant chain. As the MHC-II complex moves through the Golgi, the invariant chain is degraded, leaving the CLIP fragment in the MHC-II peptide binding groove. CLIP is eventually displaced by the loading of antigenic peptides, followed by presentation on the APC cell surface to CD4+ T-cells.
  • For Immunoprecipitation
Recognizes human CLIP. Detects a band of ~33-35kDa by Western blot.
Type: Primary
Antigen: CLIP
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: