Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Nitrotyrosine
- Antigensymbol:Nitro-Tyr
- Klonalitet:Monoclonal
- ELISA:Yes
- Värd:Mouse
- Isotyp:IgG1
- Western blot:Yes
- Seq:02
- Form:Lyophilized from 1 ml of 2× PBS containing 0,09% sodium azide, PEG, and sucrose.
- Rening:Thiophilic adsorption and size exclusion chromatography purified
- Förp.:100 µG
Specifikationer
Om denna produkt
The enzymatic conversion of arginine to citrulline by nitric oxide synthase (NO Synthase) leads to the formation of nitric oxide (NO). Peroxynitrite is formed following reaction of NO with superoxide, producing a potent oxidant that reacts with a variety of biological molecules, including tyrosine residues in proteins. Tyrosine nitration can inactivate enzymes and receptors that depend on tyrosine residues for their activity. Nitration prevents phosphorylation of tyrosine residues important for signal transduction.
- For ELISA, Flow Cytometry, Western Blot
Recognizes human, mouse, rat and dog Nitrotyrosine.
Reconstitute with 1ml water (15 minutes at room temperature).
Type: Primary
Antigen: NITROTYROSINE
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human